Assay Details
| Gene Symbol: LOC4342538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791204.2 | XP_015646690.1 | 9 - 10 | 2102 | 65 | |
| GenBank mRNA | AK062003.1 | - | 6 - 7 | 748 | 65 | |
| AK065897.1 | - | 9 - 10 | 2104 | 65 | ||
More Information
Set Membership: |
|

LOC4342538
Genomic Map