Assay Details
| Gene Symbol: LOC4342720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791177.2 | XP_015646663.1 | 1 | 557 | 118 | |
| GenBank mRNA | AK107696.1 | - | 1 | 546 | 118 | |
More Information
Set Membership: |
|

LOC4342720
Genomic Map