Assay Details
| Gene Symbol: LOC4342961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789320.2 | XP_015644806.1 | 1 - 2 | 552 | 96 | |
| GenBank mRNA | AK060569.1 | - | 1 - 2 | 487 | 96 | |
| AK073103.1 | - | 1 - 2 | 521 | 96 | ||
More Information
Set Membership: |
|

LOC4342961
Genomic Map