Assay Details
| Gene Symbol: LOC9270956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791733.2 | XP_015647219.1 | 3 - 4 | 1002 | 65 | |
| GenBank mRNA | AK120277.1 | - | 3 - 4 | 970 | 65 | |
| AK243066.1 | - | 3 - 4 | 934 | 65 | ||
More Information
Set Membership: |
|

LOC9270956
Genomic Map