Assay Details
| Gene Symbol: LOC4343565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791010.2 | XP_015646496.1 | 9 - 10 | 5049 | 119 | |
| GenBank mRNA | AK066706.1 | - | 1 - 2 | 1922 | 119 | |
| AK100968.1 | - | 1 - 2 | 1173 | 119 | ||
More Information
Set Membership: |
|

LOC4343565
Genomic Map