Assay Details
| Gene Symbol: LOC4343565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791010.2 | XP_015646496.1 | 10 - 11 | 5248 | 113 | |
| GenBank mRNA | AK066706.1 | - | 2 - 3 | 2121 | 113 | |
| AK100968.1 | - | 2 - 3 | 1372 | 113 | ||
More Information
Set Membership: |
|

LOC4343565
Genomic Map