Assay Details
| Gene Symbol: LOC4343849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792494.1 | XP_015647980.1 | 2 - 3 | 629 | 74 | |
| GenBank mRNA | AK061294.1 | - | 2 - 3 | 635 | 74 | |
More Information
Set Membership: |
|

LOC4343849
Genomic Map