Assay Details
| Gene Symbol: LOC4343852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789826.2 | XP_015645312.1 | 1 | 652 | 114 | |
| GenBank mRNA | AK068670.1 | - | 1 | 652 | 114 | |
| AK103128.1 | - | 1 | 625 | 114 | ||
More Information
Set Membership: |
|

LOC4343852
Genomic Map