Assay Details
| Gene Symbol: LOC4344055 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791851.2 | XP_015647337.1 | 1 - 2 | 144 | 110 | |
| GenBank mRNA | AK074012.1 | - | 1 - 2 | 106 | 110 | |
| AK104592.1 | - | 1 - 2 | 81 | 110 | ||
| AK104739.1 | - | 1 - 2 | 81 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791851.2 transcripts.
Set Membership: |
|

LOC4344055
Genomic Map