Assay Details
Gene Symbol: LOC4344113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792269.2 | XP_015647755.1 | 6 - 7 | 932 | 128 | |
GenBank mRNA | AK063527.1 | - | 6 - 7 | 940 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
