Assay Details
Gene Symbol: LOC4344290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791288.2 | XP_015646774.1 | 5 - 6 | 541 | 69 | |
GenBank mRNA | AK065079.1 | - | 5 - 6 | 536 | 69 | |
AK099229.1 | - | 5 - 6 | 536 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
