Assay Details
Gene Symbol: LOC4342077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK119605.1 | - | 652 | 91 |
Gene Symbol: LOC4344439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794929.2 | XP_015650415.1 | 1 | 657 | 91 | |
GenBank mRNA | AK061654.1 | - | 1 | 651 | 91 | |
AK119206.1 | - | 1 | 660 | 91 | ||
AK119709.1 | - | 1 | 650 | 91 | ||
AK120393.1 | - | 1 | 652 | 91 | ||
D30763.1 | - | 1 | 645 | 91 |
Gene Symbol: LOC9267810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK287537.1 | - | 1451 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
