Assay Details
| Gene Symbol: LOC4344663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792955.2 | XP_015648441.1 | 5 - 6 | 906 | 82 | |
| GenBank mRNA | AK101530.1 | - | 5 - 6 | 859 | 82 | |
| Gene Symbol: LOC9271247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794773.2 | XP_015650259.1 | 5 - 6 | 891 | 82 | |
More Information
Set Membership: |
|

LOC4344663
Genomic Map