Assay Details
Gene Symbol: LOC4344832 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794536.2 | XP_015650022.1 | 10 - 11 | 2948 | 74 | |
XM_015794537.2 | XP_015650023.1 | 11 - 12 | 2849 | 74 | ||
GenBank mRNA | AK120496.1 | - | 11 - 12 | 2805 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
