Assay Details
| Gene Symbol: LOC4345448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795411.2 | XP_015650897.1 | 5 - 6 | 1073 | 80 | |
| GenBank mRNA | AK065829.1 | - | 5 - 6 | 1064 | 80 | |
More Information
Set Membership: |
|

LOC4345448
Genomic Map