Assay Details
| Gene Symbol: LOC4345690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793648.2 | XP_015649134.1 | 1 - 2 | 347 | 78 | |
| GenBank mRNA | AK069725.1 | - | 1 - 2 | 354 | 78 | |
More Information
Set Membership: |
|

LOC4345690
Genomic Map