Assay Details
| Gene Symbol: LOC4345690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793648.2 | XP_015649134.1 | 3 - 4 | 579 | 72 | |
| GenBank mRNA | AK069725.1 | - | 3 - 4 | 586 | 72 | |
More Information
Set Membership: |
|

LOC4345690
Genomic Map