Assay Details
Gene Symbol: LOC4345755 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015793059.2 | XP_015648545.1 | 3 - 4 | 946 | 61 | |
GenBank mRNA | AK106955.1 | - | 3 - 4 | 861 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
