Assay Details
| Gene Symbol: LOC4340204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787431.2 | XP_015642917.1 | 4 - 5 | 937 | 58 | |
| GenBank mRNA | AK073443.1 | - | 4 - 5 | 923 | 58 | |
More Information
Set Membership: |
|

LOC4340204
Genomic Map