Assay Details
| Gene Symbol: LOC4346520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795681.2 | XP_015651167.1 | 10 - 11 | 1139 | 96 | |
| GenBank mRNA | AK059654.1 | - | 8 - 9 | 840 | 96 | |
| AK102536.1 | - | 10 - 11 | 1014 | 96 | ||
More Information
Set Membership: |
|

LOC4346520
Genomic Map