Assay Details
| Gene Symbol: LOC4346520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795681.2 | XP_015651167.1 | 14 - 15 | 1477 | 82 | |
| GenBank mRNA | AK059654.1 | - | 12 - 13 | 1178 | 82 | |
| AK102536.1 | - | 14 - 15 | 1352 | 82 | ||
More Information
Set Membership: |
|

LOC4346520
Genomic Map