Assay Details
| Gene Symbol: LOC4346520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795681.2 | XP_015651167.1 | 2 - 3 | 292 | 81 | |
| GenBank mRNA | AK102536.1 | - | 2 - 3 | 167 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795681.2 transcripts.
Set Membership: |
|

LOC4346520
Genomic Map