Assay Details
| Gene Symbol: LOC4346520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795681.2 | XP_015651167.1 | 3 - 4 | 392 | 81 | |
| GenBank mRNA | AK059654.1 | - | 1 - 2 | 93 | 81 | |
| AK102536.1 | - | 3 - 4 | 267 | 81 | ||
More Information
Set Membership: |
|

LOC4346520
Genomic Map