Assay Details
| Gene Symbol: LOC4346520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795681.2 | XP_015651167.1 | 8 - 9 | 954 | 72 | |
| GenBank mRNA | AK059654.1 | - | 6 - 7 | 655 | 72 | |
| AK102536.1 | - | 8 - 9 | 829 | 72 | ||
More Information
Set Membership: |
|

LOC4346520
Genomic Map