Assay Details
| Gene Symbol: LOC4346520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795681.2 | XP_015651167.1 | 9 - 10 | 1039 | 67 | |
| GenBank mRNA | AK059654.1 | - | 7 - 8 | 740 | 67 | |
| AK102536.1 | - | 9 - 10 | 914 | 67 | ||
More Information
Set Membership: |
|

LOC4346520
Genomic Map