Assay Details
| Gene Symbol: LOC4347086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756510.2 | XP_015611996.1 | 2 - 3 | 471 | 89 | |
| GenBank mRNA | AK065475.1 | - | 2 - 3 | 472 | 89 | |
More Information
Set Membership: |
|

LOC4347086
Genomic Map