Assay Details
| Gene Symbol: LOC4347262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756625.1 | XP_015612111.1 | 1 - 2 | 244 | 83 | |
| XM_015756626.1 | XP_015612112.1 | 1 - 2 | 244 | 83 | ||
| GenBank mRNA | AK063988.1 | - | 1 - 2 | 241 | 83 | |
| AK099330.1 | - | 1 - 2 | 242 | 83 | ||
| AK099392.1 | - | 1 - 2 | 238 | 83 | ||
More Information
Set Membership: |
|

LOC4347262
Genomic Map