Assay Details
| Gene Symbol: LOC4347262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756626.1 | XP_015612112.1 | 2 - 3 | 375 | 114 | |
| GenBank mRNA | AK063988.1 | - | 2 - 3 | 372 | 114 | |
| AK099330.1 | - | 2 - 3 | 373 | 114 | ||
| AK099392.1 | - | 2 - 3 | 369 | 114 | ||
More Information
Set Membership: |
|

LOC4347262
Genomic Map