Assay Details
Gene Symbol: LOC4347610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756807.2 | XP_015612293.1 | 13 - 14 | 1949 | 75 | |
GenBank mRNA | AK060889.1 | - | 6 - 7 | 621 | 75 | |
AK101403.1 | - | 13 - 14 | 1941 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
