Assay Details
Gene Symbol: LOC4347610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756807.2 | XP_015612293.1 | 1 - 2 | 553 | 128 | |
GenBank mRNA | AK101403.1 | - | 1 - 2 | 545 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
