Assay Details
Gene Symbol: LOC4347610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756807.2 | XP_015612293.1 | 4 - 5 | 945 | 78 | |
GenBank mRNA | AK101403.1 | - | 4 - 5 | 937 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
