Assay Details
| Gene Symbol: LOC4347745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756659.1 | XP_015612145.1 | 1 - 2 | 1259 | 97 | |
| GenBank mRNA | AK100579.1 | - | 1 - 2 | 1185 | 97 | |
More Information
Set Membership: |
|

LOC4347745
Genomic Map