Assay Details
Gene Symbol: LOC4348186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757353.2 | XP_015612839.1 | 12 - 13 | 1297 | 106 | |
GenBank mRNA | AK068119.1 | - | 12 - 13 | 1241 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
