Assay Details
| Gene Symbol: LOC4348186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757353.2 | XP_015612839.1 | 2 - 3 | 355 | 81 | |
| GenBank mRNA | AK068119.1 | - | 2 - 3 | 299 | 81 | |
More Information
Set Membership: |
|

LOC4348186
Genomic Map