Assay Details
| Gene Symbol: LOC4348767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK061298.1 | - | 1 | 838 | 120 | |
| AK070851.1 | - | 2 | 419 | 120 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4348767| Gene Symbol: LOC4348767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK061298.1 | - | 1 | 838 | 120 | |
| AK070851.1 | - | 2 | 419 | 120 | ||
Set Membership: |
|