Assay Details
| Gene Symbol: LOC4348803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757515.2 | XP_015613001.1 | 2 - 3 | 289 | 80 | |
| GenBank mRNA | AK059744.1 | - | 2 - 3 | 287 | 80 | |
| EU152202.1 | - | 2 - 3 | 243 | 80 | ||
More Information
Set Membership: |
|

LOC4348803
Genomic Map