Assay Details
Gene Symbol: LOC4348984 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758667.2 | XP_015614153.1 | 2 - 3 | 268 | 54 | |
GenBank mRNA | AK240700.1 | - | 2 - 3 | 249 | 54 |
Gene Symbol: LOC4335698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK240828.1 | - | 2156 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
