Assay Details
| Gene Symbol: LOC4349012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757504.2 | XP_015612990.1 | 1 - 2 | 412 | 53 | |
| GenBank mRNA | AK120584.1 | - | 1 - 2 | 379 | 53 | |
More Information
Set Membership: |
|

LOC4349012
Genomic Map