Assay Details
Gene Symbol: LOC4349228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757292.2 | XP_015612778.1 | 1 - 2 | 292 | 88 | |
XM_015757293.2 | XP_015612779.1 | 1 - 2 | 297 | 88 | ||
GenBank mRNA | AK071890.1 | - | 1 - 2 | 295 | 88 | |
AK101036.1 | - | 1 - 2 | 298 | 88 | ||
AK120472.1 | - | 1 - 2 | 297 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015757292.2, XM_015757293.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
