Assay Details
Gene Symbol: LOC4349228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757292.2 | XP_015612778.1 | 4 - 5 | 1516 | 149 | |
XM_015757293.2 | XP_015612779.1 | 4 - 5 | 1521 | 149 | ||
GenBank mRNA | AK058873.1 | - | 2 - 3 | 1054 | 149 | |
AK071890.1 | - | 4 - 5 | 1519 | 149 | ||
AK101036.1 | - | 4 - 5 | 1522 | 149 | ||
AK120472.1 | - | 4 - 5 | 1521 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
