Assay Details
Gene Symbol: LOC4349228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757292.2 | XP_015612778.1 | 6 - 7 | 1673 | 69 | |
XM_015757293.2 | XP_015612779.1 | 6 - 7 | 1678 | 69 | ||
GenBank mRNA | AK058873.1 | - | 4 - 5 | 1211 | 69 | |
AK071890.1 | - | 6 - 7 | 1676 | 69 | ||
AK101036.1 | - | 6 - 7 | 1679 | 69 | ||
AK120472.1 | - | 6 - 7 | 1678 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
