Assay Details
| Gene Symbol: LOC4349237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759313.2 | XP_015614799.1 | 6 - 7 | 982 | 71 | |
| GenBank mRNA | AK070135.1 | - | 6 - 7 | 748 | 71 | |
| AY551919.1 | - | 5 - 6 | 585 | 71 | ||
More Information
Set Membership: |
|

LOC4349237
Genomic Map