Assay Details
Gene Symbol: LOC9268613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758756.2 | XP_015614242.1 | 2 - 3 | 3212 | 63 | |
GenBank mRNA | AK064083.1 | - | 1 - 2 | 577 | 63 |
Gene Symbol: LOC107279238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759531.2 | XP_015615017.1 | 2 - 3 | 2809 | 63 |
Gene Symbol: LOC107281160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785641.2 | XP_015641127.1 | 2 - 3 | 2852 | 63 |
Gene Symbol: LOC107281487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789434.2 | XP_015644920.1 | 2 - 3 | 2881 | 63 |
Gene Symbol: LOC9267767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781348.2 | XP_015636834.1 | 2 - 3 | 3867 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
