Assay Details
Gene Symbol: LOC4351289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764590.2 | XP_015620076.1 | 13 - 14 | 2335 | 120 | |
XM_015764591.2 | XP_015620077.1 | 13 - 14 | 2334 | 120 | ||
GenBank mRNA | AY491431.1 | - | 12 - 13 | 1645 | 120 |
Gene Symbol: LOC9266997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761672.2 | XP_015617158.1 | 13 - 14 | 2262 | 120 | |
XM_015761673.2 | XP_015617159.1 | 13 - 14 | 2261 | 120 | ||
GenBank mRNA | AK318576.1 | - | 3 - 4 | 709 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
