Assay Details
| Gene Symbol: LOC4351289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764590.2 | XP_015620076.1 | 17 - 18 | 3576 | 99 | |
| XM_015764591.2 | XP_015620077.1 | 17 - 18 | 3572 | 99 | ||
| GenBank mRNA | AY491431.1 | - | 16 - 17 | 2883 | 99 | |
| Gene Symbol: LOC9266997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761672.2 | XP_015617158.1 | 17 - 18 | 3503 | 99 | |
| XM_015761673.2 | XP_015617159.1 | 17 - 18 | 3499 | 99 | ||
| GenBank mRNA | AK318576.1 | - | 7 - 8 | 1950 | 99 | |
More Information
Set Membership: |
|

LOC4351289
Genomic Map