Assay Details
Gene Symbol: LOC4351289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764590.2 | XP_015620076.1 | 19 - 20 | 3842 | 126 | |
XM_015764591.2 | XP_015620077.1 | 19 - 20 | 3838 | 126 |
Gene Symbol: LOC9266997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761672.2 | XP_015617158.1 | 19 - 20 | 3769 | 129 | |
XM_015761673.2 | XP_015617159.1 | 19 - 20 | 3765 | 129 | ||
GenBank mRNA | AK318576.1 | - | 9 - 10 | 2216 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
