Assay Details
Gene Symbol: LOC4351289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764590.2 | XP_015620076.1 | 20 - 21 | 3941 | 62 | |
XM_015764591.2 | XP_015620077.1 | 20 - 21 | 3937 | 62 | ||
GenBank mRNA | AY491431.1 | - | 19 - 20 | 3248 | 62 |
Gene Symbol: LOC9266997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761672.2 | XP_015617158.1 | 20 - 21 | 3871 | 62 | |
XM_015761673.2 | XP_015617159.1 | 20 - 21 | 3867 | 62 | ||
GenBank mRNA | AK318576.1 | - | 10 - 11 | 2318 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
