Assay Details
Gene Symbol: LOC4351289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764590.2 | XP_015620076.1 | 6 - 7 | 1303 | 68 | |
XM_015764591.2 | XP_015620077.1 | 6 - 7 | 1302 | 68 | ||
GenBank mRNA | AY491431.1 | - | 5 - 6 | 613 | 68 |
Gene Symbol: LOC9266997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761672.2 | XP_015617158.1 | 6 - 7 | 1230 | 68 | |
XM_015761673.2 | XP_015617159.1 | 6 - 7 | 1229 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
