Assay Details
| Gene Symbol: LOC4351575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762659.2 | XP_015618145.1 | 2 - 3 | 336 | 81 | |
| GenBank mRNA | AK111505.1 | - | 2 - 3 | 264 | 81 | |
| DQ201200.1 | - | 1 - 2 | 128 | 81 | ||
More Information
Set Membership: |
|

LOC4351575
Genomic Map