Assay Details
Gene Symbol: LOC4351575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762659.2 | XP_015618145.1 | 8 - 9 | 837 | 107 | |
GenBank mRNA | AK111505.1 | - | 8 - 9 | 765 | 107 | |
DQ201200.1 | - | 7 - 8 | 629 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
